Learn More
Trajan Scientific HPLC Kit MyCapLC Medical 3/PK
SDP
Supplier:  Trajan Scientific 2251111
Encompass_Preferred
MyCapLC kit containing components to assemble three complete 1/16 OD x 150 m ID x 50 mm length UHPLC columns with 250 mm inlet and 100 mm outlet tails 1/32 OD x 50 m ID
Catalog No. 50-004-94087