Learn More
Trajan Scientific HPLC Kit MyCapLC Medical 3/PK
Supplier: Trajan Scientific 2251110
MyCapLC kit containing components to assemble three complete 1/16 OD x 150 m ID x 50 mm length UHPLC columns with 250 mm inlet and 100 mm outlet tails 360 m OD x 50 m ID