Learn More
Trajan Scientific HPLC Kit MyCapLC Medical 3/PK
SDP
Catalog No. 5000494627
Encompass_Preferred
MyCapLC kit containing components to assemble three complete 1/16 OD x 300 m ID x 50 mm length UHPLC columns with 250 mm inlet and 100 mm outlet tails 1/32 OD x 50 m ID
Catalog No. 50-004-94627 Supplier Trajan Scientific Supplier No. 2251131
MyCapLC kit containing components to assemble three complete 1/16 OD x 300 m ID x 50 mm length UHPLC columns with 250 mm inlet and 100 mm outlet tails 1/32 OD x 50 m ID