Learn More
Trajan Scientific HPLC Kit MyCapLC Medical 3/PK
SDP
Supplier:  Trajan Scientific 2251331
Encompass_Preferred
MyCapLC kit containing components to assemble three complete 1/16 OD x 300 m ID x 150 mm length UHPLC columns with 250 mm inlet and 100 mm outlet tails 1/32 OD x 50 m ID
Catalog No. 50-004-93784