Learn More
Waters Corp Waters Corporation HPLC Melamine Analysis Package, for Melamine Screening, with Oasis Cartridges, Low ppb Detection
Catalog No. 50780058 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
These analysis packages are designed for the screening of melamine and melamine-related compounds in foods, including infant formulas and dairy products. They incorporate Oasis cartridges, vials, filters, and a selection of LC columns (either HPLC or UPLC). Optimized to deliver rapid and accurate results at low ppb levels, these packages assist in meeting the challenges of food testing.
  • Designed for screening of melamine and melamine-related compounds in foods, including infant formulas and dairy products.
  • Incorporates Oasis cartridges (as specified in US FDA Laboratory Information Bulletin [LIB] No. 4422).
  • Includes vials, filters, and a selection of LC columns (HPLC or UPLC).
  • Optimized for quick and accurate results down to low ppb levels.
  • Aids in meeting the challenges of food testing.
Catalog No. 50-780-058 Supplier Waters Corp Supplier No. 176001773
These analysis packages are designed for the screening of melamine and melamine-related compounds in foods, including infant formulas and dairy products. They incorporate Oasis cartridges, vials, filters, and a selection of LC columns (either HPLC or UPLC). Optimized to deliver rapid and accurate results at low ppb levels, these packages assist in meeting the challenges of food testing.
  • Designed for screening of melamine and melamine-related compounds in foods, including infant formulas and dairy products.
  • Incorporates Oasis cartridges (as specified in US FDA Laboratory Information Bulletin [LIB] No. 4422).
  • Includes vials, filters, and a selection of LC columns (HPLC or UPLC).
  • Optimized for quick and accurate results down to low ppb levels.
  • Aids in meeting the challenges of food testing.