Learn More
Trajan Scientific HPLC PROTECOL Amino Guard 1/EA
SDP
Catalog No. 5000494198
Encompass_Preferred
Amino x 5 m particle size x 120 pore size 250 mm length x 4 6 mm ID stainless steel with Guard ProteCol HPLC column
Catalog No. 50-004-94198 Supplier Trajan Scientific Supplier No. 2AM05R46SG
Amino x 5 m particle size x 120 pore size 250 mm length x 4 6 mm ID stainless steel with Guard ProteCol HPLC column