Learn More
Trajan Scientific HPLC PROTECOL C18 Medical 1/EA
SDP
Catalog No. 5000494154
Encompass_Preferred
C18G x 3 m particle size x 200 pore size 150 mm length x 150 m ID PEEKsil ProteCol HPLC capillary column
Catalog No. 50-004-94154 Supplier Trajan Scientific Supplier No. 2C18103N15K
C18G x 3 m particle size x 200 pore size 150 mm length x 150 m ID PEEKsil ProteCol HPLC capillary column