Learn More
Trajan Scientific HPLC PROTECOL C18 Medical 1/EA
SDP
Catalog No. 5000494156
Encompass_Preferred
C18G x 3 m particle size x 200 pore size 250 mm length x 300 m ID stainless steel ProteCol HPLC column with custom tails
Catalog No. 50-004-94156 Supplier Trajan Scientific Supplier No. 2C18103R30KSC
C18G x 3 m particle size x 200 pore size 250 mm length x 300 m ID stainless steel ProteCol HPLC column with custom tails