Learn More
Trajan Scientific HPLC PROTECOL C18 Medical 1/EA
SDP
Catalog No. 5000494625
Encompass_Preferred
C18G x 5 m particle size x 300 pore size 150 mm length x 150 m ID
Catalog No. 50-004-94625 Supplier Trajan Scientific Supplier No. 2C18403N15K
C18G x 5 m particle size x 300 pore size 150 mm length x 150 m ID