Learn More
Trajan Scientific HPLC PROTECOL C18 Medical 1/EA
SDP
Catalog No. 5000494694
Encompass_Preferred
C18G x 3 m particle size x 200 pore size 250 mm length x 300 m ID stainless steel ProteCol HPLC column
Catalog No. 50-004-94694 Supplier Trajan Scientific Supplier No. 2C18103R30K
Add to Cart
Add to Cart
C18G x 3 m particle size x 200 pore size 250 mm length x 300 m ID stainless steel ProteCol HPLC column