Learn More
Trajan Scientific HPLC PROTECOL C18 Medical 3/PK
SDP
Catalog No. 5000493703
Encompass_Preferred
C18G x 3 m particle size x 300 pore size 10 mm length x 530 m ID (IT450 OT30) PEEKsil ProteCol HPLC guard column PK3
Catalog No. 50-004-93703 Supplier Trajan Scientific Supplier No. 2C18403G53K01
C18G x 3 m particle size x 300 pore size 10 mm length x 530 m ID (IT450 OT30) PEEKsil ProteCol HPLC guard column PK3