Learn More
Trajan Scientific HPLC PROTECOL C8 Medical 1/EA
SDP
Catalog No. 5000493692
Encompass_Preferred
C8H x 3 m particle size x 1000 pore size 100 mm length x 150 m ID PEEKsil ProteCol HPLC capillary column
Catalog No. 50-004-93692 Supplier Trajan Scientific Supplier No. 2C8203M15K
C8H x 3 m particle size x 1000 pore size 100 mm length x 150 m ID PEEKsil ProteCol HPLC capillary column