Learn More
Trajan Scientific HPLC PROTECOL C8 Medical 1/EA
SDP
Catalog No. 5000494153
Encompass_Preferred
C8H x 3 m particle size x 1000 pore size 150 mm length x 150 m ID PEEKsil ProteCol HPLC capillary column
Catalog No. 50-004-94153 Supplier Trajan Scientific Supplier No. 2C8203N15K
C8H x 3 m particle size x 1000 pore size 150 mm length x 150 m ID PEEKsil ProteCol HPLC capillary column