Learn More
Trajan Scientific HPLC PROTECOL C8 Medical 3/PK
SDP
Catalog No. 5000494161
Encompass_Preferred
C8H x 3 m particle size x 1000 pore size 10 mm length x 150 m ID PEEKsil ProteCol HPLC trap column
Catalog No. 50-004-94161 Supplier Trajan Scientific Supplier No. 2C8203T15K
C8H x 3 m particle size x 1000 pore size 10 mm length x 150 m ID PEEKsil ProteCol HPLC trap column