Learn More
Trajan Scientific HPLC PROTECOL C8 Medical 3/PK
SDP
Catalog No. 5000493845
Encompass_Preferred
C8Q 10 mm length x 530 m ID ProteCol HPLC guard column PK3
Catalog No. 50-004-93845 Supplier Trajan Scientific Supplier No. 2C8403G53K
C8Q 10 mm length x 530 m ID ProteCol HPLC guard column PK3