Learn More
Trajan Scientific HPLC PROTECOL Polar Medical 3/PK
SDP
Catalog No. 5000493857
Encompass_Preferred
C18P x 3 m particle size x 120 pore size 10 mm length x 300 m ID PEEKsil ProteCol HPLC trap column
Catalog No. 50-004-93857 Supplier Trajan Scientific Supplier No. 2POL03T30K
Add to Cart
Add to Cart
C18P x 3 m particle size x 120 pore size 10 mm length x 300 m ID PEEKsil ProteCol HPLC trap column