Learn More
Trajan Scientific HPLC Protecol UHPLC Column 1/EA
SDP
Catalog No. 5000494338
Encompass
30 mm length x 1 0 mm ID ProteCol bio-inert UHPLC glass lined column 10-32 thread 0 5 m frit
Catalog No. 50-004-94338 Supplier Trajan Scientific Supplier No. 08291541
30 mm length x 1 0 mm ID ProteCol bio-inert UHPLC glass lined column 10-32 thread 0 5 m frit