Learn More
Trajan Scientific HPLC Protecol UHPLC Fitting 2/PK
SDP
Catalog No. 5000494236
Encompass
10-32 end fittings for ProteCol bio-inert UHPLC 2 1 mm ID columns PK2
Catalog No. 50-004-94236 Supplier Trajan Scientific Supplier No. 08290000
10-32 end fittings for ProteCol bio-inert UHPLC 2 1 mm ID columns PK2