Learn More
Trajan Scientific HPLC Protecol UHPLC Fitting 2/PK
SDP
Catalog No. 5000494237
Encompass
6-32 end fittings for ProteCol bio-inert UHPLC 2 1 mm ID columns PK2
Catalog No. 50-004-94237 Supplier Trajan Scientific Supplier No. 08290001
6-32 end fittings for ProteCol bio-inert UHPLC 2 1 mm ID columns PK2