Learn More
Trajan Scientific HPLC Protecol UHPLC Fitting 2/PK
SDP
Catalog No. 5000494398
Encompass
6-32 end fittings for ProteCol bio-inert UHPLC 1 0 mm ID columns PK2
Catalog No. 50-004-94398 Supplier Trajan Scientific Supplier No. 08290003
6-32 end fittings for ProteCol bio-inert UHPLC 1 0 mm ID columns PK2