Learn More
Trajan Scientific HPLC Protecol UHPLC Tube 1/EA
SDP
Catalog No. 5000494337
Encompass
250 mm length x 1 0 mm ID ProteCol bio-inert UHPLC glass lined column tubing
Catalog No. 50-004-94337 Supplier Trajan Scientific Supplier No. 08290140
250 mm length x 1 0 mm ID ProteCol bio-inert UHPLC glass lined column tubing