Learn More
Trajan Scientific HPLC Protecol UHPLC Tube 1/EA
SDP
Supplier:  Trajan Scientific 08290340
Encompass
100 mm length x 1 0 mm ID ProteCol bio-inert UHPLC glass lined column tubing
Catalog No. 50-004-93760