Learn More
Trajan Scientific HPLC Reverse Phase Protecol C18 1/EA
SDP
Catalog No. 5000494159
Encompass_Preferred
C18Q x 3 m particle size x 100 pore size 150 mm length x 4 6 mm ID PEEK coated with guard ProteCol HPLC column
Catalog No. 50-004-94159 Supplier Trajan Scientific Supplier No. 2C18303N46PG
C18Q x 3 m particle size x 100 pore size 150 mm length x 4 6 mm ID PEEK coated with guard ProteCol HPLC column