Learn More
Trajan Scientific HPLC Reverse Phase Protecol C18 1/EA
SDP
Catalog No. 5000494160
Encompass_Preferred
C18Q x 5 m particle size x 100 pore size 150 mm length x 4 6 mm ID stainless steel ProteCol HPLC column
Catalog No. 50-004-94160 Supplier Trajan Scientific Supplier No. 2C18305N46S
C18Q x 5 m particle size x 100 pore size 150 mm length x 4 6 mm ID stainless steel ProteCol HPLC column