Learn More
Trajan Scientific HPLC Reverse Phase Protecol C18 1/EA
SDP
Catalog No. 5000494123
Encompass_Preferred
C18G x 3 m particle size x 120 pore size 150 mm length x 4 6 mm ID stainless steel ProteCol HPLC column
Catalog No. 50-004-94123 Supplier Trajan Scientific Supplier No. 2C18503N46S
C18G x 3 m particle size x 120 pore size 150 mm length x 4 6 mm ID stainless steel ProteCol HPLC column