Learn More
Trajan Scientific HPLC Reverse Phase Protecol C18 1/EA
SDP
Catalog No. 5000494056
Encompass_Preferred
C18H x 5 m particle size x 120 pore size 150 mm length x 2 1 mm ID PEEK coated ProteCol HPLC column
Catalog No. 50-004-94056 Supplier Trajan Scientific Supplier No. 2C18203N02P
C18H x 5 m particle size x 120 pore size 150 mm length x 2 1 mm ID PEEK coated ProteCol HPLC column