Learn More
Trajan Scientific HPLC Reverse Phase Protecol C18 1/EA
SDP
Catalog No. 5000494058
Encompass_Preferred
C18H x 5 m particle size x 120 pore size 150 mm length x 3 mm ID stainless steel ProteCol HPLC column
Catalog No. 50-004-94058 Supplier Trajan Scientific Supplier No. 2C18203N03S
C18H x 5 m particle size x 120 pore size 150 mm length x 3 mm ID stainless steel ProteCol HPLC column