Learn More
Trajan Scientific HPLC Reverse Phase Protecol C18 1/EA
SDP
Catalog No. 5000494624
Encompass_Preferred
C18G x 2 5 m particle size x 120 pore size 50 mm length x 2 1 mm ID stainless steel ProteCol HPLC column
Catalog No. 50-004-94624 Supplier Trajan Scientific Supplier No. 2C18525L02S
C18G x 2 5 m particle size x 120 pore size 50 mm length x 2 1 mm ID stainless steel ProteCol HPLC column