Learn More
Trajan Scientific HPLC Reverse Phase Protecol C18 1/EA
SDP
Catalog No. 5000494003
Encompass_Preferred
C18G x 5 m particle size x 120 pore size 250 mm length x 4 6 mm ID stainless steel ProteCol HPLC column
Catalog No. 50-004-94003 Supplier Trajan Scientific Supplier No. 2C18505R46S
C18G x 5 m particle size x 120 pore size 250 mm length x 4 6 mm ID stainless steel ProteCol HPLC column