Learn More
Trajan Scientific HPLC Reverse Phase Protecol C18 1/EA
SDP
Catalog No. 5000493822
Encompass_Preferred
C18Q x 3 m particle size x 100 pore size 50 mm length x 4 6 mm ID PEEK coated with guard ProteCol HPLC column
Catalog No. 50-004-93822 Supplier Trajan Scientific Supplier No. 2C18303L46PG
C18Q x 3 m particle size x 100 pore size 50 mm length x 4 6 mm ID PEEK coated with guard ProteCol HPLC column