Learn More
Trajan Scientific HPLC Reverse Phase Protecol C18 1/EA
SDP
Catalog No. 5000493780
Encompass_Preferred
C18G x 5 m particle size x 120 pore size 250 mm length x 4 6 mm ID PEEK coated ProteCol HPLC column
Catalog No. 50-004-93780 Supplier Trajan Scientific Supplier No. 2C18505R46P
C18G x 5 m particle size x 120 pore size 250 mm length x 4 6 mm ID PEEK coated ProteCol HPLC column