Learn More
Trajan Scientific HPLC Reverse Phase Protecol C18 1/EA
SDP
Catalog No. 5000493808
Encompass_Preferred
C18G x 3 m particle size x 200 pore size 150 mm length x 2 1 mm ID PEEK coated ProteCol HPLC column
Catalog No. 50-004-93808 Supplier Trajan Scientific Supplier No. 2C18103N02P
C18G x 3 m particle size x 200 pore size 150 mm length x 2 1 mm ID PEEK coated ProteCol HPLC column