Learn More
Trajan Scientific HPLC Reverse Phase Protecol C18 1/EA
SDP
Catalog No. 5000493811
Encompass_Preferred
C18Q x 5 m particle size x 100 pore size 250 mm length x 2 1 mm ID PEEK coated ProteCol HPLC column
Catalog No. 50-004-93811 Supplier Trajan Scientific Supplier No. 2C18305R02P
C18Q x 5 m particle size x 100 pore size 250 mm length x 2 1 mm ID PEEK coated ProteCol HPLC column