Learn More
Trajan Scientific HPLC Reverse Phase Protecol C18 1/EA
SDP
Catalog No. 5000493690
Encompass_Preferred
C18Q x 5 m particle size x 100 pore size 150 mm length x 4 6 mm ID PEEK coated ProteCol HPLC column
Catalog No. 50-004-93690 Supplier Trajan Scientific Supplier No. 2C18305N46P
C18Q x 5 m particle size x 100 pore size 150 mm length x 4 6 mm ID PEEK coated ProteCol HPLC column