Learn More
Trajan Scientific HPLC Reverse Phase Protecol C18 1/EA
SDP
Catalog No. 5000493686
Encompass_Preferred
C18H x 5 m particle size x 120 pore size 150 mm length x 4 6 mm ID PEEK coated ProteCol HPLC column
Catalog No. 50-004-93686 Supplier Trajan Scientific Supplier No. 2C18205N46P
C18H x 5 m particle size x 120 pore size 150 mm length x 4 6 mm ID PEEK coated ProteCol HPLC column