Learn More
Trajan Scientific HPLC Reverse Phase Protecol C18 1/EA
SDP
Catalog No. 5000494200
Encompass_Preferred
C18Q x 3 m particle size x 100 pore size 150 mm length x 2 1 mm ID PEEK coated ProteCol HPLC column
Catalog No. 50-004-94200 Supplier Trajan Scientific Supplier No. 2C18303N02P
C18Q x 3 m particle size x 100 pore size 150 mm length x 2 1 mm ID PEEK coated ProteCol HPLC column