Learn More
Trajan Scientific HPLC Reverse Phase Protecol C18 1/EA
SDP
Catalog No. 5000494695
Encompass_Preferred
C18Q x 3 m particle size x 100 pore size 50 mm length x 2 1 mm ID stainless steel with guard ProteCol HPLC column
Catalog No. 50-004-94695 Supplier Trajan Scientific Supplier No. 2C18303L02SG
C18Q x 3 m particle size x 100 pore size 50 mm length x 2 1 mm ID stainless steel with guard ProteCol HPLC column