Learn More
Trajan Scientific HPLC Reverse Phase Protecol C18 1/EA
SDP
Catalog No. 5000494045
Encompass_Preferred
C18G x 3 m particle size x 120 pore size 50 mm length x 4 6 mm ID PEEK coated ProteCol HPLC column
Catalog No. 50-004-94045 Supplier Trajan Scientific Supplier No. 2C18503L46P
C18G x 3 m particle size x 120 pore size 50 mm length x 4 6 mm ID PEEK coated ProteCol HPLC column