Learn More
Trajan Scientific HPLC Reverse Phase Protecol C18 1/EA
SDP
Catalog No. 5000494048
Encompass_Preferred
C18G x 3 m particle size x 120 pore size 100 mm length x 4 6 mm ID stainless steel with guard ProteCol HPLC column
Catalog No. 50-004-94048 Supplier Trajan Scientific Supplier No. 2C18503M46SG
C18G x 3 m particle size x 120 pore size 100 mm length x 4 6 mm ID stainless steel with guard ProteCol HPLC column