Learn More
Trajan Scientific HPLC Reverse Phase Protecol C18 3/PK
SDP
Catalog No. 5000494121
Encompass_Preferred
C18G x 3 m particle size x 300 pore size 10 mm length x 300 m ID PEEKsil ProteCol HPLC trap column
Catalog No. 50-004-94121 Supplier Trajan Scientific Supplier No. 2C18403T30K
C18G x 3 m particle size x 300 pore size 10 mm length x 300 m ID PEEKsil ProteCol HPLC trap column