Learn More
Trajan Scientific HPLC Reverse Phase Protecol C18 3/PK
SDP
Catalog No. 5000493781
Encompass_Preferred
C18G x 5 m particle size x 120 pore size 10 mm length x 4 0 mm ID stainless steel PK3 ProteCol HPLC guard column PK3
Catalog No. 50-004-93781 Supplier Trajan Scientific Supplier No. 2C18505G04S
C18G x 5 m particle size x 120 pore size 10 mm length x 4 0 mm ID stainless steel PK3 ProteCol HPLC guard column PK3