Learn More
Trajan Scientific HPLC Reverse Phase Protecol C18 3/PK
SDP
Catalog No. 5000493803
Encompass_Preferred
C18G x 3 m particle size x 200 pore size 10 mm length x 2 1 mm ID stainless steel PK3 ProteCol HPLC guard column PK3
Catalog No. 50-004-93803 Supplier Trajan Scientific Supplier No. 2C18103G02S
C18G x 3 m particle size x 200 pore size 10 mm length x 2 1 mm ID stainless steel PK3 ProteCol HPLC guard column PK3