Learn More
Trajan Scientific HPLC Reverse Phase Protecol C18 3/PK
SDP
Catalog No. 5000493809
Encompass_Preferred
C18H x 3 m particle size x 120 pore size 10 mm length x 2 1 mm ID stainless steel ProteCol HPLC guard column PK3
Catalog No. 50-004-93809 Supplier Trajan Scientific Supplier No. 2C18203G02S
C18H x 3 m particle size x 120 pore size 10 mm length x 2 1 mm ID stainless steel ProteCol HPLC guard column PK3