Learn More
Trajan Scientific HPLC Reverse Phase Protecol C18 3/PK
SDP
Catalog No. 5000494092
Encompass_Preferred
C18Q x 5 m particle size x 100 pore size 10 mm length x 4 0 mm ID stainless steel ProteCol HPLC guard column PK3
Catalog No. 50-004-94092 Supplier Trajan Scientific Supplier No. 2C18305G04S
C18Q x 5 m particle size x 100 pore size 10 mm length x 4 0 mm ID stainless steel ProteCol HPLC guard column PK3