Learn More
Trajan Scientific HPLC Reverse Phase Protecol C8 1/EA
SDP
Catalog No. 5000494696
Encompass_Preferred
C8H x 3 m particle size x 1000 pore size 150 mm length x 4 6 mm ID PEEK coated ProteCol HPLC column
Catalog No. 50-004-94696 Supplier Trajan Scientific Supplier No. 2C8203N46P
C8H x 3 m particle size x 1000 pore size 150 mm length x 4 6 mm ID PEEK coated ProteCol HPLC column