Learn More
Trajan Scientific HPLC Reverse Phase Protecol C8 1/EA
SDP
Catalog No. 5000494701
Encompass_Preferred
C8H x 3 m particle size x 120 pore size 150 mm length x 2 1 mm ID PEEK coated ProteCol HPLC column
Catalog No. 50-004-94701 Supplier Trajan Scientific Supplier No. 2C8303N02P
C8H x 3 m particle size x 120 pore size 150 mm length x 2 1 mm ID PEEK coated ProteCol HPLC column