Learn More
Trajan Scientific HPLC Reverse Phase Protecol C8 1/EA
SDP
Catalog No. 5000493850
Encompass_Preferred
C8H x 5 m particle size x 120 pore size 150 mm length x 4 6 mm ID stainless steel with guard ProteCol HPLC column
Catalog No. 50-004-93850 Supplier Trajan Scientific Supplier No. 2C8305N46SG
C8H x 5 m particle size x 120 pore size 150 mm length x 4 6 mm ID stainless steel with guard ProteCol HPLC column