Learn More
Trajan Scientific HPLC Reverse Phase Protecol C8 3/PK
SDP
Catalog No. 5000494000
Encompass_Preferred
C8Hx 5 m particle size x 120 pore size 10 mm length x 2 1 mm ID stainless steel PK3 ProteCol HPLC guard column PK3
Catalog No. 50-004-94000 Supplier Trajan Scientific Supplier No. 2C8305G02S
C8Hx 5 m particle size x 120 pore size 10 mm length x 2 1 mm ID stainless steel PK3 ProteCol HPLC guard column PK3