Learn More
Trajan Scientific HPLC Reverse Phase Protecol C8 3/PK
SDP
Catalog No. 5000493853
Encompass_Preferred
C8A 10 mm length x 300 m ID (IT450 OT30) ProteCol HPLC trap column PK3
Catalog No. 50-004-93853 Supplier Trajan Scientific Supplier No. 2C8403T30K
Add to Cart
Add to Cart
C8A 10 mm length x 300 m ID (IT450 OT30) ProteCol HPLC trap column PK3