Learn More
Trajan Scientific HPLC Reverse Phase Protecol Polar 1/EA
SDP
Catalog No. 5000494143
Encompass_Preferred
C18P x 3 m particle size x 120 pore size 150 mm length x 4 6 mm ID PEEK coated ProteCol HPLC column
Catalog No. 50-004-94143 Supplier Trajan Scientific Supplier No. 2POL03N46P
C18P x 3 m particle size x 120 pore size 150 mm length x 4 6 mm ID PEEK coated ProteCol HPLC column